missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-17A/F Heterodimer Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Molekulargewicht 14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions
Name IL-17A/F Heterodimer
Menge 25μg
Quelle E. coli-derived mouse IL-17A/F Heterodimer protein, (N-terminus) Mouse IL-17A (Thr22-Ala158) Accession &Num; Q62386 Mouse IL-17F (Arg21-Ala153,with an N-terminal Met) Accession &Num; AAM77568 (C-terminus)
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse IL-17A/F Heterodimer Protein is derived from E. coli. The Recombinant Mouse IL-17A/F Heterodimer Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
IL-17A/F Heterodimer
E. coli-derived mouse IL-17A/F Heterodimer protein, (N-terminus) Mouse IL-17A (Thr22-Ala158) Accession &Num; Q62386 Mouse IL-17F (Arg21-Ala153,with an N-terminal Met) Accession &Num; AAM77568 (C-terminus)
Disulfide-linked heterodimer
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen