missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-17A/F Heterodimer Recombinant Protein

Marke:  R&D Systems™ 5390-IL-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415424

  • 283.81 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
IL-17A/F Heterodimer
25μg
Disulfide-linked heterodimer
Yes
14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Videos