missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-1ra/IL-1F3 Recombinant Protein

Marke:  R&D Systems™ 480-RM-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415104

  • 1046.99 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Arg8-Gln159, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Citrate, NaCl, EDTA and DTT.
Entrez Gene ID: 16181
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Sodium Citrate, NaCl, EDTA and DTT with BSA as a carrier protein.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16181
IL-1ra/IL-1F3
50μg
Unlabeled
Videos