missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-1ra/IL-1F3 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Gen-ID (Entrez) 16181
Molekulargewicht 17 kDa
Name IL-1ra/IL-1F3
Rekombinant Yes
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse IL-1ra/IL-1F3 Protein is derived from E. coli. The Recombinant Mouse IL-1ra/IL-1F3 Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
17 kDa
Yes
16181
IL-1ra/IL-1F3
E. coli-derived mouse IL-1ra/IL-1F3 protein Arg8-Gln159,with an N-terminal Met
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen