missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-4 Recombinant Protein

Marke:  R&D Systems™ 404-ML-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414044

  • 899.79 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, His23-Ser140, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16189
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16189
IL-4
50μg
Unlabeled
Videos