missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-9 Recombinant Protein

Marke:  R&D Systems™ 409-ML-010

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414144

  • 300.97 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 23-01-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gln19-Pro144
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16198
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16198
IL-9
10μg
Unlabeled
Videos