missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Lefty-1 Recombinant Protein

Marke:  R&D Systems™ 994-LF-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416504

  • 333.97 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Leu22-Arg135 (pro) & Leu136-Pro368 (mature), with an N-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 13590
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
26.9 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
13590
Lefty-1
25μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos