missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Neurturin Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Marke:  R&D Systems™ 477-MN-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415084

  • 481.02 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 01-08-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Recombinant Mouse Neurturin Protein is derived from E. coli. The Recombinant Mouse Neurturin Protein has been validated for the following applications: Bioactivity, Binding Activity.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose.
11.7 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Disulfide-linked homodimer
Yes
18188
Neurturin
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos