missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse NT-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
N.V.
Spezifikation
Molekulargewicht | 14 kDa |
---|---|
Name | NT-4 |
Menge | 5μg |
Quelle | Mouse myeloma cell line,NS0-derived mouse NT-4 protein Gly80-Ala209 |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Beschreibung
The Recombinant Mouse NT-4 Protein is derived from NS0. The Recombinant Mouse NT-4 Protein has been validated for the following applications: Bioactivity.Spezifikation
14 kDa | |
5μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
NT-4 | |
Mouse myeloma cell line,NS0-derived mouse NT-4 protein Gly80-Ala209 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |