missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse NT-4 Recombinant Protein

Marke:  R&D Systems™ 3236-N4-005/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412624

  • 344.53 € / 5 Mikrogramm


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured in a cell proliferation assay using BaF mouse pro-B cells transfected with TrkB. The ED 50 for this effect is 2-8 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
NT-4
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
14 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Videos