missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Oncostatin M/OSM Recombinant Protein

Marke:  R&D Systems™ 495-MO-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415214

  • 389.41 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured in a cell proliferation assay using NIH-3T3 mouse embryonic fibroblast cells. The ED 50 for this effect is 0.25-1 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
21 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
18413
Oncostatin M/OSM
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos