missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Oncostatin M/OSM Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Gen-ID (Entrez) 18413
Molekulargewicht 21 kDa
Name Oncostatin M/OSM
Menge 25μg
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse Oncostatin M (OSM) Protein is derived from E. coli. The Recombinant Mouse Oncostatin M (OSM) Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
21 kDa
25μg
E. coli-derived mouse Oncostatin M/OSM protein Ala24-Arg206,with an N-terminal Met
18413
Oncostatin M/OSM
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen