missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse PlGF-2 Recombinant Protein

Marke:  R&D Systems™ 465-PL-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415024

  • 295.69 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala24-Pro158 & Ala27-Pro158
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 18654
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
15.1 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
18654
PlGF-2
10μg
Disulfide-linked homodimer
Yes
Videos