missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse PLTP Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Gen-ID (Entrez) 18830
Molekulargewicht 53.5 kDa
Name PLTP
Menge 25μg
Quelle Mouse myeloma cell line,NS0-derived mouse PLTP protein Glu18-Ala493,with a C-terminal 6-His tag
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse PLTP Protein is derived from NS0. The Recombinant Mouse PLTP Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

18830
PLTP
Mouse myeloma cell line,NS0-derived mouse PLTP protein Glu18-Ala493,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
53.5 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen