missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse PLTP Recombinant Protein

Marke:  R&D Systems™ 4918-PL-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415184

  • 353.77 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Glu18-Ala493, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 53.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 18830
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
53.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
18830
PLTP
25μg
Unlabeled
Videos