missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse R-Spondin 3 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Gen-ID (Entrez) 72780
Molekulargewicht 28.9 kDa
Name R-Spondin 3
Menge 25μg
Quelle Chinese Hamster Ovary cell line,CHO-derived mouse R-Spondin 3 protein Met1-His277
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse R-Spondin 3 Protein is derived from CHO. The Recombinant Mouse R-Spondin 3 Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

72780
R-Spondin 3
Chinese Hamster Ovary cell line,CHO-derived mouse R-Spondin 3 protein Met1-His277
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
28.9 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen