missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse SCF/c-kit Ligand Recombinant Protein

Marke:  R&D Systems™ 455-MC-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414934

  • 1274.82 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Lys26-Ala189, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 17311
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
18.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
17311
SCF/c-kit Ligand
50μg
Unlabeled
Videos