missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse SCF Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Gen-ID (Entrez) 17311
Molekulargewicht 18.6 kDa
Name SCF/c-kit Ligand
Quelle E. coli-derived mouse SCF/c-kit Ligand protein Lys26-Ala189,with an N-terminal Met
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse SCF Protein is derived from E. coli. The Recombinant Mouse SCF Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

17311
SCF/c-kit Ligand
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
18.6 kDa
E. coli-derived mouse SCF/c-kit Ligand protein Lys26-Ala189,with an N-terminal Met
c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen