missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TNF-alpha aa 80-235 Recombinant Protein, N-terminal Met

Marke:  R&D Systems™ 410-MT-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414164

  • 285.13 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 28-01-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Leu80-Leu235, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21926
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
21926
TNF-alpha aa 80-235
10μg
A small amount of recombinant protein lacking the N-terminal methionine and four additional amino acid residues may be also present.
Yes
Videos