missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TNF RI/TNFRSF1A Fc Chimera Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Gen-ID (Entrez) 21937
Molekulargewicht 48.6 kDa (monomer)
Name TNF RI/TNFRSF1A Fc Chimera
Menge 50μg
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse TNF RI/TNFRSF1A Fc Chimera Protein is derived from NS0. The Recombinant Mouse TNF RI/TNFRSF1A Fc Chimera Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
48.6 kDa (monomer)
50μg
Mouse myeloma cell line,NS0-derived mouse TNF RI/TNFRSF1A protein, (N-terminus) Mouse TNF RI (Leu30-Ala212) Accession &Num; P25118 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
21937
TNF RI/TNFRSF1A Fc Chimera
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen