missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TNF RI/TNFRSF1A Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 430-RI-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414544

  • 403.02 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 48.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21937
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
48.6 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
21937
TNF RI/TNFRSF1A Fc Chimera
50μg
Disulfide-linked homodimer
Yes
Videos