missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TRAIL/TNFSF10 Recombinant Protein

Marke:  R&D Systems™ 1121-TL-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418603

  • 431.62 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Pro118-Asn291
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Tris, (NH4)2SO4 and ZnSO4.
Entrez Gene ID: 22035
Regulatory Status: RUO

Spezifikation

Spezifikation

Supplied as a 0.2μm filtered solution in Tris, Ammonium Sulfate and ZnSO4.
20 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
22035
TRAIL/TNFSF10
10μg
<0.01 EU per 1μg of the protein by the LAL method.
Yes
Videos