missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TSG Recombinant Protein

Marke:  R&D Systems™ 756-TG-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416244

  • 403.02 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 23-01-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Cys25-Phe222, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 23.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 65960
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
23.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
65960
TSG
50μg
Unlabeled
Videos