missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TSLP Recombinant Protein

Marke:  R&D Systems™ 555-TS-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415494

  • 494.02 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse IL-7 R alpha . The ED 50 for this effect is 0.5-2.5 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
15 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
<0.1 EU per 1μg of the protein by the LAL method.
Yes
53603
TSLP
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos