missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TSLP Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Gen-ID (Entrez) 53603
Molekulargewicht 15 kDa
Name TSLP
Menge 10μg
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse TSLP Protein is derived from NS0. The Recombinant Mouse TSLP Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
15 kDa
10μg
Mouse myeloma cell line,NS0-derived mouse TSLP protein Tyr20-Glu140,with a C-terminal 10-His tag
53603
TSLP
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen