missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse VCAM-1/CD106 Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 643-VM-200

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415804

  • 959.95 € / 200 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Phe25-Glu698
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 101.9 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22329
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
101.9 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived Met Mouse VCAM-1 (Phe25-Glu698) Accession # CAA47989 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag
<0.1 EU per 1μg of the protein by the LAL method.
Yes
22329
VCAM-1/CD106 Fc Chimera
200μg
Disulfide-linked homodimer
Unlabeled
Videos