missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Wnt-4 Recombinant Protein

Marke:  R&D Systems™ 475-WN-005

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415074

  • 481.02 € / 5 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser23-Arg351
Purity: 60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 22417
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
36.6 kDa
60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
22417
Wnt-4
5μg
Unlabeled
Videos