missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Wnt-9b Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Gen-ID (Entrez) 22412
Molekulargewicht 36.8 kDa
Name Wnt-9b
Menge 25μg
Quelle Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-9b protein Ala24-Arg359
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse Wnt-9b Protein is derived from CHO. The Recombinant Mouse Wnt-9b Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

22412
Wnt-9b
Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-9b protein Ala24-Arg359
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
36.8 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution.
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen