missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rabbit IL-4 Recombinant Protein

Marke:  R&D Systems™ 6939-RB-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415994

  • 481.02 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Arg25-Ser147, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 100302454
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rabbit
Yes
100302454
IL-4
25μg
Unlabeled
Videos