missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat CCL27/CTACK Recombinant Protein

Marke:  R&D Systems™ 4434-CK-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414754

  • 333.97 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Leu70-Lys164
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 362505
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
10.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
362505
CCL27/CTACK
25μg
Unlabeled
Videos