missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat CCL3/MIP-1 alpha Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

N.V.

Spezifikation

Gen-ID (Entrez) 25542
Molekulargewicht 7.8 kDa
Name CCL3/MIP-1 alpha
Menge 25μg
Quelle E. coli-derived rat CCL3/MIP-1 alpha protein Ala24-Ala92
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Rat CCL3/MIP-1 alpha Protein is derived from E. coli. The Recombinant Rat CCL3/MIP-1 alpha Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

25542
CCL3/MIP-1 alpha
E. coli-derived rat CCL3/MIP-1 alpha protein Ala24-Ala92
Monomer
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
7.8 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen