missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat CCL3/MIP-1 alpha Recombinant Protein

Marke:  R&D Systems™ 6625-MA-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415874

  • 195.36 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ala24-Ala92
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25542
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
7.8 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
25542
CCL3/MIP-1 alpha
25μg
Monomer
Yes
Videos