missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat CXCL3/CINC-2 alpha Recombinant Protein

Marke:  R&D Systems™ 516-CA-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415324

  • 295.69 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Arg33-Ser100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 171551
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
171551
CXCL3/CINC-2 alpha
25μg
Unlabeled
Videos