missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat GDNF Recombinant Protein

Marke:  R&D Systems™ 512-GF-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415304

  • 847.32 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ser78-Ile211
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 25453
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
15 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
25453
GDNF
50μg
Disulfide-linked homodimer
Yes
Videos