missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-2 Recombinant Protein

Marke:  R&D Systems™ 502-RL-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415254

  • 602.85 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ala21-Gln155, with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol.
Entrez Gene ID: 116562
Regulatory Status: RUO

Spezifikation

Spezifikation

Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol.
16 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
116562
IL-2
50μg
Unlabeled
Videos