missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
N.V.
Spezifikation
Gen-ID (Entrez) | 116562 |
---|---|
Molekulargewicht | 16 kDa |
Name | IL-2 |
Quelle | E. coli-derived rat IL-2 protein Ala21-Gln155,with and without an N-terminal Met |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
Beschreibung
The Recombinant Rat IL-2 Protein is derived from E. coli. The Recombinant Rat IL-2 Protein has been validated for the following applications: Bioactivity.Spezifikation
116562 | |
IL-2 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. | |
Recombinant |
16 kDa | |
E. coli-derived rat IL-2 protein Ala21-Gln155,with and without an N-terminal Met | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |