Learn More
R&D Systems™ Rhesus Macaque IL-5 Recombinant Protein
Marke:  R&D Systems™ 2707-RM-025/CF
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411774
  • 481.02 € / 25 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED 50 for this effect is typically 0.1-0.6 ng/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS.
IL-5
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
13.1 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rhesus Macaque
Disulfide-linked homodimer