missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Viral CCI Fc Chimera (CHO-expressed) Recombinant Protein

Marke:  R&D Systems™ 7055-CC-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416044

  • 333.97 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-Val258
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 52.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
CCI Fc Chimera (CHO-expressed)
25μg
Disulfide-linked homodimer
Yes
52.6 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line, CHO-derived Viral CCI (Met1-Val258) Accession # P19063 IEGRMD Human IgG1 (Pro100-Lys330)
Unlabeled
Videos