missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-1,2-Bis(2,4,6-Trimethylphenyl)-1,2-Ethandiamindihydrochlorid, 95 %, ee 99 %, Thermo Scientific™

Catalog Number p-7028597
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1301706-47-7
C20H30Cl2N2
369.374
MFCD06797063
HSFKFAWYBTVLDG-WUMQWIPTSA-N
r,r-bis-mesityl ethylenediamine dihydrochloride, r,r-1,2-bis 2,4,6-trimethylphenyl-1,2-ethanediamine dihydrochloride, 1r,2r-1,2-bis 2,4,6-trimethylphenyl ethane-1,2-diamine dihydrochloride, r,r-1,2-bis 2,4,6-trimethylphenyl-1,2-ethanediamine dihydrochloride, ee
45925672
(1R,2R)-1,2-bis(2,4,6-trimethylphenyl)ethan-1,2-diamin;Dihydrochlorid
CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C.Cl.Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1301706-47-7
C20H30Cl2N2
369.374
MFCD06797063
HSFKFAWYBTVLDG-WUMQWIPTSA-N
r,r-bis-mesityl ethylenediamine dihydrochloride, r,r-1,2-bis 2,4,6-trimethylphenyl-1,2-ethanediamine dihydrochloride, 1r,2r-1,2-bis 2,4,6-trimethylphenyl ethane-1,2-diamine dihydrochloride, r,r-1,2-bis 2,4,6-trimethylphenyl-1,2-ethanediamine dihydrochloride, ee
45925672
(1R,2R)-1,2-bis(2,4,6-trimethylphenyl)ethan-1,2-diamin;Dihydrochlorid
CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C.Cl.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (R,R)-1,2-Bis(2,4,6-trimethylphenyl)-1,2-ethanediamine dihydrochloride
Hinweise zum Prozentgehalt ee 99 %
Menge 250 mg
Formelmasse 369.38
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'