Learn More
(R,S)-Stylopin, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Tetrahydrocoptisine |
| CAS | 4312-32-7 |
| Summenformel | C19 H17 N O4 |
| Menge | 100 mg |
| Synonym | 4H-Bis[1,3]benzodioxolo[5,6-a:4',5'-g]quinolizine','6,7,12b,13-tetrahydro-, Berbine','2,3:9,10-bis(methylenedioxy)-','(±)- (8CI), 6,7,12b,13-Tetrahydro-4H-bis[1,3]benzodioxolo[5,6-a:4',5'-g]quinolizine, (RS)-Stylopine, (±)-Stylopine, (±)-Tetrahydrocoptisine, 2,3:9,10-Bis(methylenedioxy)berbine, Chelidamine, NSC 110382, NSC 404529, Tetrahydrocoptisine, dl-Stylopine, dl-Tetrahydrocoptisine |
| InChI-Formel | InChI=1S/C19H17NO4/c1-2-16-19(24-10-21-16)14-8-20-4-3-12-6-17-18(23-9-22-17)7-13(12)15(20)5-11(1)14/h1-2,6-7,15H,3-5,8-10H2 |
| SMILES | C1=C2CC3C4=CC5=C(C=C4CCN3CC2=C6C(=C1)OCO6)OCO5 |
| Molekulargewicht (g/mol) | 323.34 |
| Formelmasse | 323.1158 |