missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB17 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17501-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase.Spezifikation
RAB17 | |
Polyclonal | |
Unconjugated | |
RAB17 | |
RAB17, Ras related protein Rab 17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64284 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H0T7 | |
RAB17 | |
RAB17 Fusion Protein Ag11688 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |