missing translation for 'onlineSavingsMsg'
Learn More

Rab20 Antibody (OTI1F1), Alexa Fluor™ 594, Novus Biologicals™

Artikelnummer. 18151817 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18151817 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18151817 Supplier Novus Biologicals Supplier No. NBP273768AF594

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rab20 Monoclonal specifically detects Rab20 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab20
Anwendungen Western Blot
Klassifikation Monoclonal
Klon OTI1F1
Konjugat Alexa Fluor 594
Verdünnung Western Blot
Zusammensetzung 50 mM sodium borate with 0.05% sodium azide
Gen-Alias FLJ20429, RAB20, member RAS oncogene family, ras-related protein Rab-20
Gensymbole RAB20
Wirtsspezies Mouse
Immunogen Full length recombinant protein of human Rab20 (NP_060287) produced in HEK293T cells.
Menge 0.1 mL
Primär oder sekundär Primary
Gen-ID (Entrez) 55647
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.