missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RAB8A (Human) Recombinant Protein
Human RAB8A full-length ORF ( AAH02977, 1 a.a. - 207 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00004218-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MAKTYDYLFKLLLIGDSGVGKTCVLFRFSEDAFNSTFISTIGIDFKIRTIELDGKRIKLQIWDTAGQERFRTITTAYYRGAMGIMLVYDITNEKSFDNIRNWIRNIEEHASADVEKMILGNKCDVNDKRQVSKERGEKLALDYGIKFMETSAKANINVENAFFTLARDIKAKMDKKLEGNSPQGSNQGVKITPDQQKRSSFFRCVLL
Spezifikation
AAH02977 | |
RAB8A, member RAS oncogene family | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RAB8A | |
GST |
4218 | |
Wheat germ expression system | |
25 μg | |
MEL, RAB8 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |