missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG (H+L) Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen PA128546
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-28546 detects Bovine IgG from bovine samples.
Bovine IgG (H+L) Polyclonal antibody specifically detects Bovine IgG (H+L) in Bovine samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
Bovine IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
IgG fraction | |
RUO | |
Bovine | |
Liquid |
Immunohistochemistry, Western Blot | |
2 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
Bovine IgG, whole molecule | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |