Learn More
Rabbit anti-Chicken IgY (H+L) Secondary Antibody, HRP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen SA19509
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
SA1-9509 reacts with the heavy and light chains of the immunoglobulin molecule. This antibody shows minimal cross-reactivity with IgG of mammalian species SA1-9509 immunogen is immunoglobulin isolated from chicken serum. 2-8oC as an undiluted liquid for several weeks. After, dilution, do not use for more than one day. Store -20oC for up to one year.
Chicken IgY (H+L) Polyclonal antibody specifically detects Chicken IgY (H+L) in Chicken samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
Chicken IgY (H+L) | |
Polyclonal | |
HRP | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chicken | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 15mg/mL BSA, 25% glycerol and no preservative; pH 7.6 | |
Immunoglobulin isolated from chicken serum. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |