missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Chicken IgY (H+L) Secondary Antibody, Rhodamine, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen SA19512
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts with the heavy and light chains of the immunoglobulin molecule. SA1-9512 immunogen is immunoglobulin isolated from chicken serum.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Chicken IgY (H+L) | |
Polyclonal | |
Rhodamine | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chicken | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
1 mg/mL | |
PBS with 15mg/mL BSA, 25% glycerol and 0.05% sodium azide | |
Immunoglobulin isolated from chicken serum. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |