Learn More
F2RL3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of F2RL3.
Marke: Abnova PAB26190.50ug
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Coagulation factor II (thrombin) receptor-like 3 (F2RL3) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL3 is also a member of the protease-activated receptor family. F2RL3 is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. F2RL3 is activated by thrombin and trypsin. [provided by RefSeq
Spezifikation
F2RL3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of F2RL3. | |
In PBS (0.09% sodium azide) | |
Q96RI0 | |
F2RL3 | |
A synthetic peptide corresponding to 16 amino acid at N-terminus of human F2RL3. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-13 ug/mL) The optimal working dilution should be determined by the end user. | |
F2RL3 | |
PAR4 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
9002 | |
Human | |
Liquid |
For Research Use Only