Learn More
Rabbit anti-Goat IgG F(ab')2 Secondary Antibody, HRP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen 31403
Weitere Details : Netto-Gewicht : 0.00150kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot, and ICC applications. Antibody Specificity: This antibody reacts with the light chains on goat IgG and with those common to other goat immunoglobulins, based on electrophoresis. The antibody does not react with the Fc portion of goat IgG. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 2-8 °C until opened. Restore with 1.5 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at -70 °C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Goat IgG F(ab')2 Polyclonal antibody specifically detects Goat IgG F(ab')2 in Goat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
Goat IgG F(ab')2 | |
Polyclonal | |
HRP | |
Rabbit | |
1.5 mL | |
Secondary | |
4° C | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 15mg/mL BSA and no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Lyophilized |