missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Goat IgG (H+L) Secondary Antibody, AP, Invitrogen™

Rabbit Polyclonal Secondary Antibody

Marke:  Invitrogen 31300

199.34 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.00100kg

Artikelnummer. 10280014

  • 299.00 € / 1 Milliliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31300 has been successfully used in Western blot applications. Product # 31300 reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.0 mL of distilled water (0.6 mg/mL after restoration). Country of Origin: USA

Goat IgG (H+L) Polyclonal antibody specifically detects Goat IgG (H+L) in Goat samples. It is validated for Western Blot.
Spezifikation

Spezifikation

Goat IgG (H+L)
Polyclonal
Alkaline Phosphatase (AP)
Rabbit
1 mL
Secondary
4° C
IgG
Western Blot
0.6 mg/mL
0.01M tris HCl with 0.25M NaCl, 15mg/mL BSA and no preservative; pH 7.8
Antigen affinity chromatography
RUO
Goat
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote