Learn More
Rabbit anti-Goat IgG (H+L), Superclonal™ Recombinant Secondary Antibody, Invitrogen™
Rabbit Recombinant Polyclonal Secondary Antibody
Marke: Invitrogen A27011
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Goat IgG (H+L) Recombinant Polyclonal antibody specifically detects Goat IgG (H+L) in Goat samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
Goat IgG (H+L) | |
Recombinant Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Goat | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Recombinant full-length protein | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |